Merck
  • Downregulation of TTF1 in the rat hypothalamic ARC or AVPV nucleus inhibits Kiss1 and GnRH expression, leading to puberty delay.
Downregulation of TTF1 in the rat hypothalamic ARC or AVPV nucleus inhibits Kiss1 and GnRH expression, leading to puberty delay.
Reproductive biology and endocrinology : RB&E (2021-02-25)
Shaolian Zang, Xiaoqin Yin, Pin Li
ABSTRACT
TTF1 is a transcription factor that is expressed in the hypothalamus after birth and plays crucial roles in pubertal development. TTF1 may regulate the expression of the Kiss1 gene, which may drive puberty onset in the hypothalamic arcuate (ARC) and anterior ventral paraventricular (AVPV) nuclei. A dual-luciferase reporter assay was used to detect binding between TTF1 and the Kiss1 gene promoter. To investigate the effects of TTF1, we modified TTF1 expression in cell lines and in the ARC or AVPV nucleus of 21-day-old female rats via lentivirus infection. TTF1 and other puberty onset-related genes were detected by qRT-PCR and western blot analyses. The in vitro data indicated that TTF1 knockdown (KD) significantly reduced Kiss1 and GnRH expression. Overexpression (OE) of TTF1 promoted Kiss1 expression. In vivo, the expression of Kiss1 and GnRH decreased significantly in the rats with hypothalamic ARC- or AVPV-specific TTF1 KD. The TTF1-KD rats showed vaginal opening delay. H&E staining revealed that the corpus luteum was obviously reduced at the early puberty and adult stages in the rats with ARC- or AVPV-specific TTF1 KD. TTF1 bound to the promoter of the Kiss1 gene and enhanced its expression. For 21-day-old female rats, decreased TTF1 in the hypothalamic ARC or AVPV nucleus resulted in delayed vaginal opening and ovarian abnormalities. These observations suggested that TTF1 regulates puberty onset by promoting the expression of Kiss1 and plays an important role in gonad development.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human TTF1