Merck
  • Memory CD8+ T cell heterogeneity is primarily driven by pathogen-specific cues and additionally shaped by the tissue environment.
Memory CD8+ T cell heterogeneity is primarily driven by pathogen-specific cues and additionally shaped by the tissue environment.
iScience (2021-01-19)
Esmé T I van der Gracht, Guillaume Beyrend, Tamim Abdelaal, Iris N Pardieck, Thomas H Wesselink, Floortje J van Haften, Suzanne van Duikeren, Frits Koning, Ramon Arens
ABSTRACT
Factors that govern the complex formation of memory T cells are not completely understood. A better understanding of the development of memory T cell heterogeneity is however required to enhance vaccination and immunotherapy approaches. Here we examined the impact of pathogen- and tissue-specific cues on memory CD8+ T cell heterogeneity using high-dimensional single-cell mass cytometry and a tailored bioinformatics pipeline. We identified distinct populations of pathogen-specific CD8+ T cells that uniquely connected to a specific pathogen or associated to multiple types of acute and persistent infections. In addition, the tissue environment shaped the memory CD8+ T cell heterogeneity, albeit to a lesser extent than infection. The programming of memory CD8+ T cell differentiation during acute infection is eventually superseded by persistent infection. Thus, the plethora of distinct memory CD8+ T cell subsets that arise upon infection is dominantly sculpted by the pathogen-specific cues and further shaped by the tissue environment.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Desossiribonucleasi I, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Collagenasi, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid