Merck
  • Progress towards the development of non-peptide orally-active gonadotropin-releasing hormone (GnRH) antagonists: therapeutic implications.
Progress towards the development of non-peptide orally-active gonadotropin-releasing hormone (GnRH) antagonists: therapeutic implications.
British medical bulletin (2001-03-20)
R P Millar, Y F Zhu, C Chen, R S Struthers
ABSTRACT
Gonadotropin-releasing hormone (GnRH) is a decapeptide (pGlu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly.NH2) which is produced from a precursor polypeptide in hypothalamic neurons and secreted in a pulsatile fashion to stimulate the secretion of LH and FSH via its interaction with a cognate receptor on gonadotropes. Low doses of the native peptide delivered in a pulsatile manner to mimic that found in the hypothalamic portal vessels restore fertility in hypogonadal patients, and are also effective in treating cryptorchidism and delayed puberty. Administration of high doses of GnRH, or agonist analogues, causes desensitization of the gonadotrope with consequent decline in gonadal gametogenesis and steroid and peptide hormone synthesis. This phenomenon finds extensive therapeutic application in clinical medicine in a wide spectrum of disease (Table 1). In addition, GnRH analogues have promise as new generation male and female contraceptives in conjunction with steroid hormone replacement. GnRH antagonists inhibit the reproductive system through competition with endogenous GnRH for the receptor and, in view of their rapid effects, are being increasingly used for the above mentioned applications. The peptide agonists and antagonists currently available require parenteral administration, typically in the form of long-acting depots. A new generation of non-peptide GnRH antagonists are beginning to emerge which should allow oral administration and, therefore, may provide greater flexibility of dosing, lower costs and increased patient acceptance.