Merck
p300 Serine 89: A Critical Signaling Integrator and Its Effects on Intestinal Homeostasis and Repair.
Cancers (2021-04-04)
Keane K Y Lai, Xiaohui Hu, Keisuke Chosa, Cu Nguyen, David P Lin, Keith K Lai, Nobuo Kato, Yusuke Higuchi, Sarah K Highlander, Elizabeth Melendez, Yoshihiro Eriguchi, Patrick T Fueger, Andre J Ouellette, Nyam-Osor Chimge, Masaya Ono, Michael Kahn
ABSTRACT
Differential usage of Kat3 coactivators, CBP and p300, by β-catenin is a fundamental regulatory mechanism in stem cell maintenance and initiation of differentiation and repair. Based upon our earlier pharmacologic studies, p300 serine 89 (S89) is critical for controlling differential coactivator usage by β-catenin via post-translational phosphorylation in stem/progenitor populations, and appears to be a target for a number of kinase cascades. To further investigate mechanisms of signal integration effected by this domain, we generated p300 S89A knock-in mice. We show that S89A mice are extremely sensitive to intestinal insult resulting in colitis, which is known to significantly increase the risk of developing colorectal cancer. We demonstrate cell intrinsic differences, and microbiome compositional differences and differential immune responses, in intestine of S89A versus wild type mice. Genomic and proteomic analyses reveal pathway differences, including lipid metabolism, oxidative stress response, mitochondrial function and oxidative phosphorylation. The diverse effects on fundamental processes including epithelial differentiation, metabolism, immune response and microbiome colonization, all brought about by a single amino acid modification S89A, highlights the critical role of this region in p300 as a signaling nexus and the rationale for conservation of this residue and surrounding region for hundreds of million years of vertebrate evolution.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Roche
Cocktail di inibitori di proteasi cOmplete, Mini, senza EDTA, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Protease Inhibitor Cocktail Set V, EDTA-Free, A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases.