Merck
  • Monitoring of compound resting membrane potentials of cell cultures with ratiometric genetically encoded voltage indicators.
Monitoring of compound resting membrane potentials of cell cultures with ratiometric genetically encoded voltage indicators.
Communications biology (2021-10-09)
Philipp Rühl, Johanna M Langner, Jasmin Reidel, Roland Schönherr, Toshinori Hoshi, Stefan H Heinemann
ABSTRACT
The cellular resting membrane potential (Vm) not only determines electrical responsiveness of excitable cells but also plays pivotal roles in non-excitable cells, mediating membrane transport, cell-cycle progression, and tumorigenesis. Studying these processes requires estimation of Vm, ideally over long periods of time. Here, we introduce two ratiometric genetically encoded Vm indicators, rArc and rASAP, and imaging and analysis procedures for measuring differences in average resting Vm between cell groups. We investigated the influence of ectopic expression of K+ channels and their disease-causing mutations involved in Andersen-Tawil (Kir2.1) and Temple-Baraitser (KV10.1) syndrome on median resting Vm of HEK293T cells. Real-time long-term monitoring of Vm changes allowed to estimate a 40-50 min latency from induction of transcription to functional Kir2.1 channels in HEK293T cells. The presented methodology is readily implemented with standard fluorescence microscopes and offers deeper insights into the role of the resting Vm in health and disease.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Gramicidin from Bacillus aneurinolyticus (Bacillus brevis), Linear polypeptide antibiotic complex. A mixture of gramicidins A, B, C, and D.