Merck
A modified protocol for rapid DNA isolation from cotton (Gossypium spp.).
MethodsX (2019-02-23)
Qasim Ali, Ibrahim Bala Salisu, Ali Raza, Ahmad Ali Shahid, Abdul Qayyum Rao, Tayyab Husnain
ABSTRACT
Extraction of high-quality DNA from Gossypium (Cotton) species is notoriously difficult due to high contents of polysaccharides, quinones and polyphenols other secondary metabolites. Here, we describe a simple, rapid and modified procedure for high-quality DNA extraction from cotton, which is amenable for downstream analyses. In contrast to other CTAB methods, the described procedure is rapid, omits the use of liquid nitrogen, phenol, CsCl gradient ultracentrifugation, uses inexpensive and less hazardous reagents, and requires only ordinary laboratory equipment. The procedure employed the high concentration of NaCl and use of PVP-10 to rid the problems associated with polysaccharides and polyphenols, respectively. The average yield was approximately 10-15 μg of good quality DNA from 100 mg of tissue weight, which is adequate for projects, like genetic mapping or marker-assisted plant breeding. This protocol can be performed in as little as 3 h and may be adapted to high-throughput DNA isolation. •Buffers A and B were redesigned from Paterson et al. (1993) and Porebski et al. (1997), respectively.•Ribonuclease A was added before chloroform extraction.•A simple, rapid and inexpensive DNA extraction method is described.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Millipore
Acido etilendiamminotetraacetico