Merck
  • Covalent Immobilization of Antibodies through Tetrazine-TCO Reaction to Improve Sensitivity of ELISA Technique.
Covalent Immobilization of Antibodies through Tetrazine-TCO Reaction to Improve Sensitivity of ELISA Technique.
Biosensors (2021-12-24)
Tania García-Maceira, Fé I García-Maceira, José A González-Reyes, Luis A Torres-Sánchez, Ana Belén Aragón-Gómez, María Eugenia García-Rubiño, Elier Paz-Rojas
ABSTRACT
Enzyme-linked immunosorbent assay (ELISA) is routinely used to detect biomolecules related to several diseases facilitating diagnosis and monitoring of these, as well as the possibility of decreasing their mortality rate. Several methods have been carried out to improve the ELISA sensitivity through antibodies immobilization on the microtiter plates. Here, we have developed a strategy of antibodies immobilization to improve the ELISA sensitivity increasing the antibody density surface through the tetrazine (Tz)-trans-cyclooctene (TCO) reaction. For this, we prepared surfaces with tetrazine groups while the captured antibody was conjugated with TCO. The tetrazine surfaces were prepared in two different ways: (1) from aminated plates and (2) from Tz-BSA-coated plates. The surfaces were evaluated using two sandwich ELISA models, one of them using the low-affinity antibody anti-c-myc as a capture antibody to detect the c-myc-GST-IL8h recombinant protein, and the other one to detect the carcinoembryonic human protein (CEA). The sensitivity increased in both surfaces treated with tetrazine in comparison with the standard unmodified surface. The c-myc-GST-IL8h detection was around 10-fold more sensible on both tetrazine surfaces, while CEA ELISA detection increased 12-fold on surfaces coated with Tz-BSA. In conclusion, we show that it is possible to improve the ELISA sensitivity using this immobilization system, where capture antibodies bond covalently to surfaces.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution