Merck
Stabilized coronavirus spike stem elicits a broadly protective antibody.
Cell reports (2021-10-29)
Ching-Lin Hsieh, Anne P Werner, Sarah R Leist, Laura J Stevens, Ester Falconer, Jory A Goldsmith, Chia-Wei Chou, Olubukola M Abiona, Ande West, Kathryn Westendorf, Krithika Muthuraman, Ethan J Fritch, Kenneth H Dinnon, Alexandra Schäfer, Mark R Denison, James D Chappell, Ralph S Baric, Barney S Graham, Kizzmekia S Corbett, Jason S McLellan
ABSTRACT
Current coronavirus (CoV) vaccines primarily target immunodominant epitopes in the S1 subunit, which are poorly conserved and susceptible to escape mutations, thus threatening vaccine efficacy. Here, we use structure-guided protein engineering to remove the S1 subunit from the Middle East respiratory syndrome (MERS)-CoV spike (S) glycoprotein and develop stabilized stem (SS) antigens. Vaccination with MERS SS elicits cross-reactive β-CoV antibody responses and protects mice against lethal MERS-CoV challenge. High-throughput screening of antibody-secreting cells from MERS SS-immunized mice led to the discovery of a panel of cross-reactive monoclonal antibodies. Among them, antibody IgG22 binds with high affinity to both MERS-CoV and severe acute respiratory syndrome (SARS)-CoV-2 S proteins, and a combination of electron microscopy and crystal structures localizes the epitope to a conserved coiled-coil region in the S2 subunit. Passive transfer of IgG22 protects mice against both MERS-CoV and SARS-CoV-2 challenge. Collectively, these results provide a proof of principle for cross-reactive CoV antibodies and inform the development of pan-CoV vaccines and therapeutic antibodies.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Albumina sierica bovina, 30%±2% in 0.85% sodium chloride, aseptically filled