Merck
  • Supporting dataset on the optimization and validation of a QuEChERS-based method for the determination of 218 pesticide residues in clay loam soil.
Supporting dataset on the optimization and validation of a QuEChERS-based method for the determination of 218 pesticide residues in clay loam soil.
Data in brief (2020-11-05)
Andrea Acosta-Dacal, Cristian Rial-Berriel, Ricardo Díaz-Díaz, Mª Del Mar Bernal Suárez, Manuel Zumbado, Luis Alberto Henríquez-Hernández, Octavio P Luzardo
ABSTRACT
The dataset presented in this article supports "Optimization and validation of a method for the simultaneous environmental monitoring of 218 pesticide residues in clay loam soil" [1]. A method based on QuEChERS (Quick, Easy, Cheap, Effective, Rugged & Safe) for the extraction of pesticide and some metabolites residues was developed. The quantification of the chemicals was performed by a combination of two complementary LC-MS/MS and GC-MS/MS analyses. Detailed optimization data of the QuEChERS extraction method is provided, including (1) salt combination, (2) acidification of the solvent (3) the amount of the selected acid (Formic Acid, FA) and (4) moisturization of the soil samples prior to extraction. In addition, all the validation data are presented, including the matrix effect, which was evaluated for each analyte using the recommended procedure.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Methiocarb sulfone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland