- Generation of three human iPSC lines from PLAN (PLA2G6-associated neurodegeneration) patients.
Generation of three human iPSC lines from PLAN (PLA2G6-associated neurodegeneration) patients.
Stem cell research (2021-06-06)
Candela Machuca, Marta Correa-Vela, Deyanira García-Navas, Alejandra Darling, Irene Villalón-García, José Antonio Sánchez-Alcázar, Belén Pérez-Dueñas, Slaven Erceg, Carmen Espinós
PMID34087982
ABSTRACT
The human iPSC cell lines, PLANFiPS1-Sv4F-1 (RCPFi004-A), PLANFiPS2-Sv4F-1 (RCPFi005-A), PLANFiPS3-Sv4F-1 RCPFi006-A), derived from dermal fibroblast from three patients suffering PLAN (PLA2G6-associated neurodegeneration; MIM 256600) caused by mutations in the PLA2G6 gene, was generated by non-integrative reprogramming technology using OCT3/4, SOX2, CMYC and KLF4 reprogramming factors. The pluripotency was assessed by immunocytochemistry and RT-PCR. Differentiation capacity was verified in vitro. This iPSC line can be further differentiated toward affected cells to better understand molecular mechanisms of disease and pathophysiology.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo monoclonale di topo anti-actina, α-muscolo liscio, clone 1A4, purified from hybridoma cell culture