Merck
  • Effects of eCG and FSH on ovarian response, recovery rate and number and quality of oocytes obtained by ovum pick-up in Holstein cows.
Effects of eCG and FSH on ovarian response, recovery rate and number and quality of oocytes obtained by ovum pick-up in Holstein cows.
Animal reproduction science (2008-02-26)
Sait Sendag, Yunus Cetin, Muhammet Alan, Klaus-Gerd Hadeler, Heiner Niemann
ABSTRACT
The goal of the present study was to compare the ovarian response, oocyte yields per animal, and the morphological quality of oocytes collected by ultrasound guided follicular aspiration from Holstein cows treated either with FSH or eCG. Twenty four normal cyclic, German Holstein cows were randomly divided into two groups. Fourteen cows received 3000 IU eCG on day-4 prior to ovum pick-up (OPU) (day 0), 2 days later (day-2), 625 microg cloprostenol was administered. On day-1 GnRH was administered i.m. and 24h later OPU (day 0) was performed. In ten cows a total dose of 500 IU follicle stimulating hormone (Pluset) was administered intramuscularly in a constant dosage for 4 days with intervals of 12h, starting on day-5. Luteolysis was induced by application of 625 microg cloprostenol on day-2. On day-1 (24h after the last FSH treatment) GnRH was administered i.m. and 24h later OPU (day 0) was performed. Ovarian follicles were visualized on the ultrasound monitor, counted and recorded. All visible antral follicles were punctured. Recovered oocytes were graded morphologically based on the cumulus investment. Average follicle number in ovaries was higher in FSH group than eCG group (p<0.05). Oocyte yields per animal did not differ between FSH and eCG groups. The proportion of grade A oocytes was higher in the FSH group in the than eCG group (p<0.05). Likewise, rate of grade C oocytes in FSH group were lower than eCG group (p<0.05). In conclusion, these results suggest that ovarian response, follicle number in ovaries and oocyte quality are affected by the type of gonadotropin and FSH is better alternative than eCG for OPU treatment.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Terreno di coltura 199, HEPES Modification, with Earle′s salts, L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture