Merck
A novel tumor-associated mucin of gastrointestinal carcinoma.
Journal of cancer research and clinical oncology (2001-09-26)
K M Hong, S J Jang, G Kong, K Y Song, J G Park, D J Kim, J H Chung, J H Lee, M K Paik, H K Chung
ABSTRACT
To identify a new tumor-associated antigen, a monoclonal antibody, SC142, was produced by immunizing mice with a stomach cancer cell line. The tumor specificity of mAb SC142 was studied by immunohistochemical staining, and the biochemical characteristics of this new gastrointestinal tumor-associated antigen were also studied. The expression of SC142-reactive antigen was investigated in various cancers by immunohistochemical staining. The SC142-reactive antigen was characterized by immunoblotting, sodium metaperiodate treatment assay, O-glycanase digestion assay, and lectin binding assay. The SC142-reactive antigen was highly expressed in 78% of gastric cancers (29/37) and 87% of colon cancers (27/31). No normal colon or stomach tissues remote from the tumor were positive for the antigen. The antibody also reacted with other tumors of epithelial origin such as lung squamous cell cancer (2/4), breast ductal cancer (2/20), bladder transitional cell carcinoma (4/6), and uterine cancer (3/16). Western blot analysis of the antigen revealed glycoprotein(s) which migrated as a smear ranging from the origin of the gel to about the 80 kDa region. The reactivity of this antigen with SC142 was reduced by sodium metaperiodate treatment or O-glycanase digestion, but not by N-glycanase, suggesting that the epitope is an O-glycan. In lectin-binding assay, this antigen reacted only with wheat germ agglutinin but not with Ricinus communis agglutinin, Datura stramonium agglutinin, and Sambucus nigra agglutinin. Our findings indicate that the antigen defined by SC142 is a tumor-associated antigen that could differentiate the gastrointestinal cancer cells from the normal cells. Therefore, SC142 may become a valuable tool for the immunohistochemical diagnosis and tumor immunoscintigraphy of the gastrointestinal cancer patients.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
O-Glycosidase from Streptococcus pneumoniae, recombinant, expressed in E. coli, buffered aqueous solution