- Identification of bifunctional GA and AG intrastrand crosslinks formed between cisplatin and DNA.
Identification of bifunctional GA and AG intrastrand crosslinks formed between cisplatin and DNA.
Journal of inorganic biochemistry (2004-12-29)
Rajesh Gupta, Jennifer L Beck, Margaret M Sheil, Stephen F Ralph
PMID15621289
ABSTRACT
A combination of enzymatic digestion and electrospray ionisation mass spectrometry (ESI-MS) was used to characterise bifunctional adducts in which cisplatin is bound to GA base sequences in 8mer and 16mer oligonucleotides that do not contain other, higher affinity binding sites. The extent of formation of bifunctional adducts with GA base sequences was significant, but less than that seen with similar oligonucleotides containing either AG or GG sequences.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Phosphodiesterase II from bovine spleen, lyophilized powder, ≥5.0 units/mg protein