Merck
[Effects of gene pta disruption on L-tryptophan fermentation].
Wei sheng wu xue bao = Acta microbiologica Sinica (2011-07-30)
Jing Huang, Jianming Shi, Qian Liu, Qingyang Xu, Xixian Xie, Tingyi Wen, Ning Chen
ABSTRACT
To study the effects of gene pta disruption on biosynthesis of L-tryptophan. The pta gene of the L-tryptophan producing strain E. coli TRTH was disrupted by Red recombination technology and a pta mutant E. coli TRTHdeltapta was constructed. Fed-batch fermentation of E. coli TRTHdeltapta was carried out in 30-Liter fermentor to investigate the biomass, L-tryptophan production, organic acid content and the concentration of NH4+, lactate, pyruvate and succinate. The metabolic flux balance model of L-tryptophan synthesis by E. coli was established. Based on this model, the practical metabolic flux distribution of E. coli and its pta mutant were determined with the linear program planted in MATLAB software. Compared with E. coli TRTH, the pta mutant was able to maintain higher growth rate at exponential phase, the final biomass and the L-tryptophan production were increased by 52.7% and 46.8% respectively. Meanwhile, the data analysis of organic acids accumulated during fed-batch culture showed that the concentration of acetate was decreased to 2.5 g/L, which was only 19.5% of that of the parental strain; as the decreased concentration of succinate, the accumulation of pyruvate and lactate was increased. The concentration of Na+, K+, PO4(3-) were consistent with E. coli TRTH during the fed-batch culture, the concentration of NH4+ was decreased by 33.2%. The metabolic flux analysis indicated that EMP pathway and TCA cycle were reduced by 7.4% and 32.2% respectively, but PP pathway was increased by 8.4% compared with E. coli TRTH during the middle and late period of the fed-batch culture. In the process of L-tryptophan fermentation, pta gene deletion in E. coli TRTH led to change in metabolic flux and acetate content, which derepressed its inhibition on cell growth and production of L-tryptophan and finally made a substantial increase of bacterial biomass and L-tryptophan production.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Phosphotransacetylase from Bacillus stearothermophilus, lyophilized powder, ≥3,000 units/mg protein (biuret)