Merck
From inulin to fructose syrups using sol-gel immobilized inulinase.
Applied biochemistry and biotechnology (2011-03-30)
Gonçalo L M Santa, Susana M S A Bernardino, Salomé Magalhães, Vanessa Mendes, Marco P C Marques, Luís P Fonseca, Pedro Fernandes
ABSTRACT
The present work aims to provide the basic characterization of sol-gel immobilized inulinase, a biocatalyst configuration yet unexploited, using as model system the hydrolysis of inulin to fructose. Porous xerogel particles with dimensions in slight excess of 10 μm were obtained, yielding an immobilization efficiency of roughly 80%. The temperature- and pH-activity profiles displayed a broader bell-shaped pattern as a result of immobilization. In the latter case, a shift of the optimal pH of 0.5 pH units was observed towards a less acidic environment. The kinetic parameters estimated from the typical Michaelis-Menten kinetics suggest that immobilization in sol-gel did not tamper with the native enzyme conformation, but on the other hand, entrapment brought along mass transfer limitations. The sol-gel biocatalyst displayed a promising operational stability, since it was used in more than 20 consecutive 24-hour batch runs without noticeable decay in product yield. The performance of sol-gel biocatalyst particles doped with magnetite roughly matched the performance of simple sol-gel particles in a single batch run. However, the operational stability of the former proved poorer, since activity decay was evident after four consecutive 24-hour batch runs.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Inulinase from Aspergillus niger, lyophilized, powder, brown-gray, ~25 U/mg