Merck
  • Association interaction and voltammetric determination of 1-aminopyrene and 1-hydroxypyrene at cyclodextrin and DNA based electrochemical sensors.
Association interaction and voltammetric determination of 1-aminopyrene and 1-hydroxypyrene at cyclodextrin and DNA based electrochemical sensors.
Bioelectrochemistry (Amsterdam, Netherlands) (2005-08-27)
A Ferancová, M Bucková, E Korgová, O Korbut, P Gründler, I Wärnmark, R Stepán, J Barek, J Zima, J Labuda
ABSTRACT
The aim of this work was voltammetric determination of 1-aminopyrene and 1-hydroxypyrene using carbon paste electrodes modified with cyclodextrin derivatives and double stranded deoxyribonucleic acid (dsDNA). The detection schemes based on a preconcentration and differential pulse voltammetric (DPV) determination at beta-cyclodextrin and gamma-cyclodextrin modified carbon paste electrode (beta-CD/CPE, gamma-CD/CPE), neutral beta-cyclodextrin polymer and carboxymethyl-beta-cyclodextrin polymer modified screen-printed electrode (beta-CDP/SPE, beta-CDPA/SPE) and dsDNA modified screen-printed electrode (DNA/SPE) are proposed for the trace determination of studied analytes within the concentration range from 2 x 10(-8) to 4 x 10(-7) mol dm(-3) and from 2 x 10(-7) to 4 x 10(-6) mol dm(-3) with the limits of quantification down to 10(-8) mol dm(-3). Depending on pH, 1-aminopyrene interacts with both surface attached CD and DNA by electrostatic bonds and supramolecular complexation while 1-hydroxypyrene associates with the CD hosts via complexation. The 1-aminopyrene interaction with dsDNA was confirmed by fluorimetric measurements in the solution phase using a competing DNA-TO-PRO-3 dye complex. In addition, the effect of temperature on this association was investigated using an electrically heated DNA-modified carbon paste electrode (DNA/CPE).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1-Aminopyrene, 97%