Merck
  • Simultaneous quantitative analysis of metabolites using ion-pair liquid chromatography-electrospray ionization mass spectrometry.
Simultaneous quantitative analysis of metabolites using ion-pair liquid chromatography-electrospray ionization mass spectrometry.
Analytical chemistry (2006-09-15)
Leon Coulier, Richard Bas, Sonja Jespersen, Elwin Verheij, Mariët J van der Werf, Thomas Hankemeier
ABSTRACT
We have developed an analytical method, consisting of ion-pair liquid chromatography coupled to electrospray ionization mass spectrometry (IP-LC-ESI-MS), for the simultaneous quantitative analysis of several key classes of polar metabolites, like nucleotides, coenzyme A esters, sugar nucleotides, and sugar bisphosphates. The use of the ion-pair agent hexylamine and optimization of the pH of the mobile phases were critical parameters in obtaining good retention and peak shapes of many of the above-mentioned polar and acidic metabolites that are impossible to analyze using standard reversed-phase LC/MS. Optimum conditions were found when using a gradient from 5 mM hexylamine in water (pH 6.3) to 90% methanol/10% 10 mM ammonium acetate (pH 8.5). The IP-LC-ESI-MS method was extensively validated by determining the linearity (R2 > 0.995), sensitivity (limit of detection 0.1-1 ng), repeatability, and reproducibility (relative standard deviation <10%). The IP-LC-ESI-MS method was shown to be a useful tool for microbial metabolomics, i.e., the comprehensive quantitative analysis of metabolites in extracts of microorganisms, and for the determination of the energy charge, i.e., the cellular energy status, as an overall quality measure for the sample workup and analytical protocols.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Hexylamine, 99%