Merck
  • Density functional and experimental studies on the FT-IR and FT-Raman spectra and structure of 2,6-diamino purine and 6-methoxy purine.
Density functional and experimental studies on the FT-IR and FT-Raman spectra and structure of 2,6-diamino purine and 6-methoxy purine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-05-01)
V Krishnakumar, R Ramasamy
ABSTRACT
FT-IR and FT-Raman spectra of 2,6-diamino purine (DAP) and 6-methoxy purine (MP) have been recorded in the regions of 4000-400cm(-1) and 3500-100cm(-1), respectively. The spectra were interpreted with the aid of normal coordinate analysis following full structure optimizations and force field calculations based on density functional theory (DFT) using standard B3LYP/6-31G* and B3LYP/6-311+G** method and basis set combinations. Normal coordinate calculations were performed with the DFT force field corrected by a recommended set of scaling factors yielding fairly good agreement between observed and calculated frequencies.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
2,6-Diaminopurine, 98%