Merck
  • Simultaneous speciation of methylmercury and butyltin species in environmental samples by headspace-stir bar sorptive extraction-thermal desorption-gas chromatography-mass spectrometry.
Simultaneous speciation of methylmercury and butyltin species in environmental samples by headspace-stir bar sorptive extraction-thermal desorption-gas chromatography-mass spectrometry.
Journal of chromatography. A (2008-02-19)
A Prieto, O Zuloaga, A Usobiaga, N Etxebarria, L A Fernández, C Marcic, A de Diego
ABSTRACT
In this work, making use of experimental designs, headspace-stir bar sorptive extraction (HS-SBSE) followed by thermal desorption (TD) coupled to a gas chromatography-mass spectrometry (GC-MS) for the simultaneous determination of mercury and tin organometallic compounds present in surface water, sediment and biological tissue was optimized. All solid samples require a previous extraction typically done with diluted HCl or KOH/methanol solutions, and the derivatization, in all the cases, of the organometallic compounds with NaBEt(4). As a consequence, the preconcentration step was carried out in a 0.1 mol L(-1) HOAc/NaOAc buffer solution, with 0.1% (m/v) of NaBEt(4), without the addition of NaCl as a salting out reagent, and with the stir bar (20 mm x 1 mm) located in the headspace (HS). In addition, the desorption step required the following conditions: 250 degrees C (desorption temperature), 15 min (desorption time), 14.1 psi (approximately 97.2 kPa) (vent pressure) and 75 mL min(-1) (vent flow). Finally, to assure the extraction of all the analytes under equilibrium, 5h are required. Inorganic mercury (Hg(2+)) and tripropyltin (TPrT) were used as internal standards to correct for variations in the extraction, derivatization and detection steps. The resulting method provides precise (4-17%) and accurate (against four certified reference materials) results in the ng L(-1) and pg g(-1) range concentrations with recoveries within 80-120% for water samples. The proposed methodology is currently applied in the speciation analysis of specimens obtained in different estuarine sites of the Basque Coast.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Butyltin trichloride, 95%