Merck
Microbial conversion of L-ascorbic acid to L-erythroascorbic acid.
Bioscience, biotechnology, and biochemistry (2009-04-09)
Daiki Kyotani, Keiko Hasegawa, Hideo Ohishi, Wenhui Wu, Liyun Wang, Keiji Hasumi
ABSTRACT
We found that a strain of Penicillium sp. effectively converted L-ascorbic acid to a five-carbon analog, which was identified as L-erythroascorbic acid based on spectroscopic analysis. The conversion was achieved by growing culture or washed mycelia, with a yield of approximately 20-30% (mol/mol). The processes for the bioconversion and purification of the product are described.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
D-(−)-Isoascorbic acid, 98%
Sigma-Aldrich
D-Isoascorbic acid, FG