Merck
  • Simultaneous determination of thiamphenicol, florfenicol and florfenicol amine in swine muscle by liquid chromatography-tandem mass spectrometry with immunoaffinity chromatography clean-up.
Simultaneous determination of thiamphenicol, florfenicol and florfenicol amine in swine muscle by liquid chromatography-tandem mass spectrometry with immunoaffinity chromatography clean-up.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-10-28)
Pengjie Luo, Xia Chen, Chunlai Liang, Hua Kuang, Liming Lu, Zhigang Jiang, Zhanhui Wang, Cun Li, Suxia Zhang, Jianzhong Shen
ABSTRACT
A rapid and sensitive liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS) method to quantify thiamphenicol (TAP), florfenicol (FF), and florfenicol amine (FFA) in swine muscle is described. An immunoaffinity chromatography (IAC) column based on polyclonal antibodies and protein A-sepharose CL 4B was used to clean-up extracted samples. IAC optimized conditions were found that allowed the IAC to be reused for selective binding of TAP, FF, and FFA. The dynamic column capacity was more than 512ng/mL of gel after being used for 15 cycles. From fortified swine muscle samples at levels of 0.4-50ng/g, the average recoveries were 85.2-98.9% with intra- and inter-day variations less than 9.8% and 12.4%, respectively. The limit of quantitation ranged from 0.4 to 4.0microg/kg.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Florfenicol amine, VETRANAL®, analytical standard