Merck
Numerical studies of the membrane fluorescent dyes dynamics in ground and excited states.
Biochimica et biophysica acta (2010-06-01)
Justyna Barucha-Kraszewska, Sebastian Kraszewski, Piotr Jurkiewicz, Christophe Ramseyer, Martin Hof
ABSTRACT
Fluorescence methods are widely used in studies of biological and model membranes. The dynamics of membrane fluorescent markers in their ground and excited electronic states and correlations with their molecular surrounding within the fully hydrated phospholipid bilayer are still not well understood. In the present work, Quantum Mechanical (QM) calculations and Molecular Dynamics (MD) simulations are used to characterize location and interactions of two membrane polarity probes (Prodan; 6-propionyl-2-dimethylaminonaphthalene and its derivative Laurdan; 2-dimethylamino-6-lauroylnaphthalene) with the dioleoylphosphatidylcholine (DOPC) lipid bilayer model. MD simulations with fluorophores in ground and excited states are found to be a useful tool to analyze the fluorescent dye dynamics and their immediate vicinity. The results of QM calculations and MD simulations are in excellent agreement with available experimental data. The calculation shows that the two amphiphilic dyes initially placed in bulk water diffuse within 10 ns towards their final location in the lipid bilayer. Analysis of solvent relaxation process in the aqueous phase occurs on the picoseconds timescale whereas it takes nanoseconds at the lipid/water interface. Four different relaxation time constants, corresponding to different relaxation processes, where observed when the dyes were embedded into the membrane.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N,N-Dimethyl-6-propionyl-2-naphthylamine, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)