Merck
Stereoselective α-aminoallylation of aldehydes with chiral tert-butanesulfinamides and allyl bromides.
The Journal of organic chemistry (2010-08-21)
José C González-Gómez, Mohamed Medjahdi, Francisco Foubelo, Miguel Yus
ABSTRACT
The combination of an aldehyde, an allylic bromide, and tert-butanesulfinamide in the presence of indium metal and titanium tetraethoxide allows straightforward access to homoallylamine derivatives in high yields and stereoselectivities. Moreover, the synthetic utility of the enantioenriched homoallylamine derived from n-decanal was illustrated in a concise synthesis of (+)-isosolenopsin. In this context, similar homoallylamines has been recently used by other groups in the synthesis of naturally occurring alkaloids.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Allile bromuro, ReagentPlus®, 99%, contains ≤1000 ppm propylene oxide as stabilizer
Sigma-Aldrich
Allile bromuro, reagent grade, 97%, contains ≤1000 ppm propylene oxide as stabilizer