Merck
  • [The influence of root excretions of germinating barley seed (Hordeum vulgare L.) on qualitative and quantitative composition of soil organic components].
[The influence of root excretions of germinating barley seed (Hordeum vulgare L.) on qualitative and quantitative composition of soil organic components].
Zhurnal obshchei biologii (2010-09-28)
O I Volkov
ABSTRACT
The data from scientific publications on excretory activity of herbs root endings were analyzed, along with the data on the role of polyvalent metals cations in stabilization of humus substances (HS) of soil organic mineral complex. On the base of the analysis a working hypothesis was proposed considering root endings influence on fractional composition of soil organic components. To detect the changes taking place in soil HS, the chromatographic fractionation method was chosen. The soil aggregates stuck to root endings of germinating barley seed were washed off, and the washouts were used as the samples for the analysis. The soil from the weighed portion was dissolved directly with extenuating concentrations of LiCl and Li2SO4 alkaline solution. The fractionation was carried out in a chromatographic column. Some changes were detected in optical density of chernozem and dark-grey forest soil leached out after 1-2 days of barley seeds germination. Besides, the experiment showed that the content of organic carbon in HS changes as well.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Lithium sulfate, ≥98.5% (titration)
Sigma-Aldrich
Lithium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium sulfate, purum p.a., ≥98.0% (T)