Merck
  • Vibrational spectroscopy (FTIR and FTRaman) investigation using ab initio (HF) and DFT (B3LYP) calculations on the structure of 3-Bromo phenol.
Vibrational spectroscopy (FTIR and FTRaman) investigation using ab initio (HF) and DFT (B3LYP) calculations on the structure of 3-Bromo phenol.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-12-28)
D Mahadevan, S Periandy, S Ramalingam
ABSTRACT
The FT-IR and FT-Raman spectra of 3-Bromo phenol (3-BP) molecule have been recorded using Bruker IFS 66V spectrometer in the range of 4000-100 cm(-1). The molecular geometry and vibrational frequencies in the ground state are calculated by using the ab initio Hartree-fock (HF) and DFT (B3LYP) methods with 6-31G (d, p) and 6-311G (d, p) basis sets. The computed values of frequencies are scaled using a suitable scale factor to yield good coherence with the observed values. Making use of the recorded data, the complete vibrational assignments are made and analysis of the observed fundamental bands of molecule is carried out. The geometries and normal modes of vibrations obtained from ab initio HF and B3LYP calculations are in good agreement with the experimentally observed data. The differences between the observed and scaled wave number values of most of the fundamentals are very small in DFT than HF. The inductive effect of halogen atom in the molecule has also been investigated.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
3-Bromophenol, 98%