Merck
  • Synthesis, Cu(II) complexation, 64Cu-labeling and biological evaluation of cross-bridged cyclam chelators with phosphonate pendant arms.
Synthesis, Cu(II) complexation, 64Cu-labeling and biological evaluation of cross-bridged cyclam chelators with phosphonate pendant arms.
Dalton transactions (Cambridge, England : 2003) (2011-12-16)
Riccardo Ferdani, Dannon J Stigers, Ashley L Fiamengo, Lihui Wei, Barbara T Y Li, James A Golen, Arnold L Rheingold, Gary R Weisman, Edward H Wong, Carolyn J Anderson
ABSTRACT
A new class of cross-bridged cyclam-based macrocycles featuring phosphonate pendant groups has been developed. 1,4,8,11-tetraazacyclotetradecane-1,8-di(methanephosphonic acid) (CB-TE2P, 1) and 1,4,8,11-tetraazacyclotetradecane-1-(methanephosphonic acid)-8-(methanecarboxylic acid) (CB-TE1A1P, 2) have been synthesized and have been shown to readily form neutral copper(II) complexes at room temperature as the corresponding dianions. Both complexes showed high kinetic inertness to demetallation and crystal structures confirmed complete encapsulation of copper(II) ion within each macrocycle's cleft-like structure. Unprecedented for cross-bridged cyclam derivatives, both CB-TE2P (1) and CB-TE1A1P (2) can be radiolabeled with (64)Cu at room temperature in less than 1 h with specific activities >1 mCi μg(-1). The in vivo behavior of both (64)Cu-CB-TE2P and (64)Cu-CB-TE1A1P were investigated through biodistribution studies using healthy male Lewis rats. Both new compounds showed rapid clearance with similar or lower accumulation in non-target organs/tissues when compared to other copper chelators including CB-TE2A, NOTA and Diamsar.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,4,8,11-Tetraazacyclotetradecane, 98% (GC)