Merck
  • Methylselenocysteine treatment leads to diselenide formation in human cancer cells: evidence from X-ray absorption spectroscopy studies.
Methylselenocysteine treatment leads to diselenide formation in human cancer cells: evidence from X-ray absorption spectroscopy studies.
Biochemistry (2012-01-17)
Claire M Weekley, Jade B Aitken, Ian F Musgrave, Hugh H Harris
ABSTRACT
The selenoamino acids methylselenocysteine (MeSeCys) and selenomethionine (SeMet) have disparate efficacies as anticancer agents. Herein, we use X-ray absorption spectroscopy to determine the chemical form of selenium in human neuroblastoma cells. Cells treated with MeSeCys contain a significant diselenide component, which is absent from SeMet-treated cells and suggests that metabolites of MeSeCys are capable of altering the redox status of the cells. The differences in the speciation of Se in the selenoamino acid-treated cells may provide insight into the differing anticancer activities of MeSeCys and SeMet.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Diphenyl diselenide, 98%
Sigma-Aldrich
Diphenyl diselenide, purum, ≥97.0% (GC)