Merck
Oxazolidinone-promoted, torquoselective Nazarov cyclizations.
Organic letters (2012-03-30)
Daniel J Kerr, Michael Miletic, Jason H Chaplin, Jonathan M White, Bernard L Flynn
ABSTRACT
Oxazolidinones are powerful promoters of the Nazarov reaction, enabling the cyclization of conventionally resistant substrates to be achieved under mild conditions. They exert excellent regio- and torquoselective control in both the conventional Nazarov reaction giving cyclopentenones and in the "interrupted" Nazarov reaction, giving more highly substituted multistereocenter containing products.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
2-Cyclopenten-1-one, 98%