Merck
Salidroside protects PC12 cells from MPP⁺-induced apoptosis via activation of the PI3K/Akt pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-06-06)
Lingling Zhang, Wenjun Ding, Huixing Sun, Qiong Zhou, Jingqun Huang, Xuefen Li, Yonghong Xie, Jianzong Chen
ABSTRACT
Oxidative stress plays an important role in the pathogenesis of Parkinson's disease (PD). Salidroside (SAL), a phenylpropanoid glycoside isolated from Rhodiola rosea L., can exert potent antioxidant properties. In this study, we investigated the protective effects, and the possible mechanism of action, of SAL against 1-methyl-4-phenylpyridinium (MPP(+))-induced cell damage in rat adrenal pheochromocytoma PC12 cells. Pretreatment of PC12 cells with SAL significantly reduced the ability of MPP(+) to induce apoptosis in a dose and time-dependent manner. SAL significantly and dose-dependently inhibited MPP(+)-induced chromatin condensation and MPP(+)-induced release of lactate dehydrogenase by PC12 cells. SAL enhanced Akt phosphorylation in PC12 cells, and the protective effects of SAL against MPP(+)-induced apoptosis were abolished by LY294002, a specific inhibitor of phosphatidylinositol 3-kinase (PI3K) phosphorylation. These findings suggest that SAL prevents MPP(+)-induced apoptosis in PC12 cells, at least in part through activation of the PI3K/Akt pathway.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Salidroside, analytical standard