Merck
  • Variation in penetration of submicrometric particles through electrostatic filtering facepieces during exposure to paraffin oil aerosol.
Variation in penetration of submicrometric particles through electrostatic filtering facepieces during exposure to paraffin oil aerosol.
Journal of occupational and environmental hygiene (2012-08-07)
Carmela Plebani, Stefano Listrani, Giovanna Tranfo, Francesca Tombolini
ABSTRACT
Several studies show the increase of penetration through electrostatic filters during exposure to an aerosol flow, because of particle deposition on filter fibers. We studied the effect of increasing loads of paraffin oil aerosol on the penetration of selected particle sizes through an electrostatic filtering facepiece. FFP2 facepieces were exposed for 8 hr to a flow rate of 95.0 ± 0.5 L/min of polydisperse paraffin aerosol at 20.0 ± 0.5 mg/m(3). The penetration of bis(2-ethylhexyl)sebacate (DEHS) monodisperse neutralized aerosols, with selected particle size in the 0.03-0.40 μm range, was measured immediately prior to the start of the paraffin aerosol loading and at 1, 4, and 8 hr after the start of paraffin aerosol loading. Penetration through isopropanol-treated facepieces not oil paraffin loaded was also measured to evaluate facepiece behavior when electrostatic capture mechanisms are practically absent. During exposure to paraffin aerosol, DEHS penetration gradually increased for all aerosol sizes, and the most penetrating particle size (0.05 μm at the beginning of exposure) shifted slightly to larger diameters. After the isopropanol treatment, the higher penetration value was 0.30 μm. In addition to an increased penetration during paraffin loading at a given particle size, the relative degree of increase was greater as the particle size increased. Penetration value measured after 8 hr for 0.03-μm particles was on average 1.6 times the initial value, whereas it was about 8 times for 0.40-μm particles. This behavior, as well evidenced in the measurements of isopropanol-treated facepieces, can be attributed to the increasing action in particle capture of the electrostatic forces (Coulomb and polarization), which depend strictly on the diameter and electrical charge of neutralized aerosol particles. With reference to electrostatic filtering facepieces as personal protective equipment, results suggest the importance of complying with the manufacturer instructions when it is specified that their use has to be restricted to a single shift.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Olio di paraffina, puriss., meets analytical specification of Ph. Eur., BP, viscous liquid
Supelco
Olio di paraffina, suitable for IR spectroscopy
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, ≥97.0% (GC)
Supelco
Bis(2-ethylhexyl) sebacate, Selectophore, ≥97.0%
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, technical grade, 90%