Merck
  • Flecainide and antiarrhythmic effects in a mouse model of catecholaminergic polymorphic ventricular tachycardia.
Flecainide and antiarrhythmic effects in a mouse model of catecholaminergic polymorphic ventricular tachycardia.
Trends in cardiovascular medicine (2012-08-08)
Nian Liu, Carlo Napolitano, Luigi A Venetucci, Silvia G Priori
ABSTRACT
Recent studies have shown that flecainide may be an effective therapy to prevent life-threatening arrhythmias in patients with catecholaminergic polymorphic ventricular tachycardia. Several hypotheses have been advanced to explain the antiarrhythmic mechanism of flecainide, including Na(+) channel blockade and a direct inhibitory action on the ryanodine receptor. In this article, we review the current literature on the topic and summarize the elements of the existing debate.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Flecainide acetate salt
Supelco
(±)-Flecainide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®