Merck
  • A unique pathway for the 3-aminobutyrate starter unit from L-glutamate through β-glutamate during biosynthesis of the 24-membered macrolactam antibiotic, incednine.
A unique pathway for the 3-aminobutyrate starter unit from L-glutamate through β-glutamate during biosynthesis of the 24-membered macrolactam antibiotic, incednine.
Organic letters (2012-08-30)
Makoto Takaishi, Fumitaka Kudo, Tadashi Eguchi
ABSTRACT
Incednine is a 24-membered macrolactam antibiotic produced by Streptomyces sp. ML694-90F3. A previous study demonstrated that its unique nitrogen-containing starter unit was derived from L-glutamate. To elucidate the missing link between L-glutamate and the starter unit, deuterium labeled amino acid feeding experiments were conducted. These experiments revealed that 3-[3-(2)H]aminobutyrate and β-[2,2,4,4-(2)H(4)]glutamate were incorporated into the starter moiety. The results indicate that a novel decarboxylation of β-glutamate to give 3-aminobutyrate is involved in incednine biosynthesis.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
3-Aminobutanoic acid, 97%
Sigma-Aldrich
(S)-3-Aminobutyric acid, 97%