Merck
  • A novel colloidal gold-based lateral flow immunoassay for rapid simultaneous detection of cyromazine and melamine in foods of animal origin.
A novel colloidal gold-based lateral flow immunoassay for rapid simultaneous detection of cyromazine and melamine in foods of animal origin.
Food chemistry (2013-02-16)
Tao Le, Peifeng Yan, Jian Xu, Youjing Hao
ABSTRACT
A rapid and sensitive lateral flow immunoassay (LFIA) based on competitive format was developed and validated for simultaneous detection of cyromazine (CA) and melamine (MA) in foods of animal origin. With this method, the cut-off value for the two test lines were achieved at 25 ng/g, which was lower than the maximum residue levels (MRLs) established for CA and MA. At three fortified levels (50, 100, and 150 ng/g), the recoveries for CA and MA ranged from 73.9% to 104.2% with the relative standard deviation (RSD) less than 11.9%, based on within day and interday analysis. The lower detection limit for CA and MA in matrix sample were 0.22 ng/ml and 0.26 ng/ml, respectively, which were lower than those of published literatures. A parallel analysis of CA and MA in real samples conducted by HPLC showed comparable results to those obtained from LFIA. The results of LFIA were in good agreement with those of high performance liquid chromatography (HPLC) in the analysis of CA and MA in foods of animal origin, demonstrating the practical applicability of the developed assay in real samples. Overall, to our knowledge, this is the first report of quantitative or semi-quantitative simultaneous detection for CA and MA by immunochromatographic assay.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Melamine, 99%
Supelco
Melamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melamine, analytical standard
Supelco
Cyromazin, PESTANAL®, analytical standard
Sigma-Aldrich
N-Cyclopropyl-2,4,6-triamino-1,3,5-triazine, 97%