Merck
A rapid test for chitinase activity that uses 4-methylumbelliferyl-N-acetyl-beta-D-glucosaminide.
Applied and environmental microbiology (1987-07-01)
M O'Brien, R R Colwell
ABSTRACT
A total of 101 strains of bacteria from environmental and clinical sources, most of which were gram negative, were tested for chitobiase activity by using a filter paper spot test with 4-methylumbelliferyl-N-acetyl-beta-D-glucosaminide as the substrate. The results were compared with those obtained by a conventional plate method for chitinase activity by using colloidal chitin as the substrate. There was excellent agreement in the results for both methods. The filter paper spot test with 4-methylumbelliferyl-N-acetyl-beta-D-glucosaminide has the advantages of being rapid, simple to perform, and inexpensive. This method should be adaptable to a wider range of microorganisms, particularly those with unusual growth requirements.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
4-Methylumbelliferyl N-acetyl-β-D-glucosaminide, β-N-acetylhexosaminidase substrate
Sigma-Aldrich
4-Methylumbelliferyl N-acetyl-β-D-glucosaminide, suitable for fluorescence, ≥99.0% (TLC)