Merck
Lipoprotein lipase immobilization onto porous chitosan beads.
Biotechnology progress (1994-03-01)
K Itoyama, S Tokura, T Hayashi
ABSTRACT
Lipoprotein lipase (LPL) was covalently immobilized onto the surface of porous chitosan beads (ChB) without or with spacers of different lengths. The relative activity (RA) of the immobilized LPL was found to be high toward a small ester substrate, p-nitrophenyl laurate (pNPL). LPL immobilized with spacer gave an almost constant activity, in marked contrast with the immobilized LPL without spacer, whose activity decreased with decreasing surface concentration. Further, the RA of LPL immobilized with spacer was higher than that of LPL immobilized without a spacer. The pH, thermal, and storage stabilities of the immobilized LPL were higher than those of the free one. LPL immobilized directly to the surface of ChB without any spacer gave a higher stability than that immobilized with spacer, in spite of the lower RA. The spacer effect on RA could be explained in terms of the mobility of the immobilized LPL molecule.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
4-Nitrophenyl dodecanoate, ≥98.0% (GC)