Merck
  • GERI-BP002-A, novel inhibitor of acyl-CoA: cholesterol acyltransferase produced by Aspergillus fumigatus F93.
GERI-BP002-A, novel inhibitor of acyl-CoA: cholesterol acyltransferase produced by Aspergillus fumigatus F93.
The Journal of antibiotics (1996-01-01)
Y K Kim, H W Lee, K H Son, B M Kwon, T S Jeong, D H Lee, J Shin, Y Seo, S U Kim, S H Bok
ABSTRACT
A new inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT), designated GERI-BP002-A, was isolated from the culture broth of Aspergillus fumigatus F93 by acetone extraction, EtOAc extraction, SiO2 column chromatography and reverse phase HPLC. Spectroscopic analyses of the compound identified bis (2-hydroxy-3-tert-butyl-5-methylphenyl) methane as the structure and its molecular weight and formula to be 340 and C23H32O2, respectively. GERI-BP002-A inhibited ACAT activity by 50% at the concentration of 50 microM in an enzyme assay system using rat liver microsomes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
2,2′-Methylenebis(6-tert-butyl-4-methylphenol)