Merck
  • Fructose-1,6-bisphosphate and N-acetylcysteine attenuate the formation of advanced oxidation protein products, a new class of inflammatory mediators, in vitro.
Fructose-1,6-bisphosphate and N-acetylcysteine attenuate the formation of advanced oxidation protein products, a new class of inflammatory mediators, in vitro.
Inflammation (2012-07-11)
Guilherme Vargas Bochi, Vanessa Dorneles Torbitz, Lara Peruzzolo Cargnin, Manuela Borges Sangoi, Roberto Christ Vianna Santos, Patrícia Gomes, Rafael Noal Moresco
ABSTRACT
The accumulation of advanced oxidation protein products (AOPP) has been linked to several pathological conditions. Previous studies have identified AOPP as a novel biomarker of oxidative damage to proteins and a novel class of mediator of inflammation. The aim of this study was to determine the effects of fructose-1,6-bisphosphate (FBP) and N-acetylcysteine (NAC) as well as the synergistic effect of both treatments on the formation of AOPP in vitro. For this purpose, we incubated the human serum albumin (HSA) with various hypochlorous acid (HOCl) concentrations to produce albumin-advanced oxidation protein products (HSA-AOPP). Both FBP and NAC were capable of inhibiting the formation of HOCl-induced AOPP in a concentration-dependent manner. The synergistic effect promoted by the association of these drugs showed to be more effective than when tested alone. Thus, both FBP and NAC may be good candidates to mitigate and neutralize pro-inflammatory and pro-oxidant effects of AOPP in several diseases.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
D-Fructose 1,6-bisphosphate trisodium salt hydrate, ≥98% (TLC)