Merck
  • 3-Ureidopropionate contributes to the neuropathology of 3-ureidopropionase deficiency and severe propionic aciduria: a hypothesis.
3-Ureidopropionate contributes to the neuropathology of 3-ureidopropionase deficiency and severe propionic aciduria: a hypothesis.
Journal of neuroscience research (2001-12-18)
S Kölker, J G Okun, F Hörster, B Assmann, B Ahlemeyer, D Kohlmüller, S Exner-Camps, E Mayatepek, J Krieglstein, G F Hoffmann
ABSTRACT
3-Ureidopropionate (3-UPA) is a physiologic metabolite in pyrimidine degradation. Pathological accumulation of 3-UPA in body fluids is found in 3-ureidopropionase deficiency and severe forms of propionic aciduria. Both diseases clinically present with a severe neuropathology involving gray and white matter as well as with a dystonic dyskinetic movement disorder. To date nothing is known about the toxic nature of this metabolite. The aim of the present study was to elucidate whether 3-UPA may act as endogenous neurotoxin. Exposure of cultured chick neurons to 3-UPA induced a concentration- and time-dependent neurodegeneration. Neuronal damage was reduced by the antioxidant alpha-tocopherol and the N-methyl-D-aspartate (NMDA) receptor antagonist MK-801. In contrast, the non-NMDA receptor antagonist CNQX, the metabotropic glutamate receptor antagonist L-AP3, and succinate showed no protective effect. Furthermore, 3-UPA elicited an increased production of reactive oxygen species followed by a delayed increase in intracellular calcium concentrations. Activity measurement of single respiratory chain complexes I-V revealed an inhibition of complex V activity, but not of the electron-transferring complexes I-IV by 3-UPA. In contrast, 3-UPA did not affect the mitochondrial beta-oxidation of fatty acids. In conclusion, our results provide strong evidence that 3-UPA acts as endogenous neurotoxin via inhibition of mitochondrial energy metabolism, resulting in the initiation of secondary, energy-dependent excitotoxic mechanisms.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
3-Ureidopropionic acid, ≥98.0% (T)