Merck
  • Comparison of membrane assisted solvent extraction, stir bar sorptive extraction, and solid phase microextraction in analysis of tetramine in food.
Comparison of membrane assisted solvent extraction, stir bar sorptive extraction, and solid phase microextraction in analysis of tetramine in food.
Journal of separation science (2009-03-07)
Lowri S De Jager, Gracia A Perfetti, Gregory W Diachenko
ABSTRACT
Three environmentally friendly extraction techniques, membrane assisted solvent extraction (MASE), stir bar sorptive extraction (SBSE), and headspace solid phase microextraction (HS-SPME), were compared for the direct analysis of the highly toxic rodenticide tetramine in food. The optimized MASE method was applied to seven foods fortified with tetramine and compared to previously reported SBSE and HS-SPME results. Parameters such as the standard addition linearity (MASE (0.964-0.999), SBSE (0.966-0.999), HS-SPME (0.955-0.999)), recovery (MASE (12-86%), SBSE (36-130%), HS-SPME (50-200%)), reproducibility (MASE (3.0-30%), SBSE (4.4-9.6%), HS-SPME (1-12%)), and LOD (MASE (1.6-6.4 ng/g), SBSE (0.2-2.1 ng/g), HS-SPME (0.9-4.3 ng/g)) were compared.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Tetramethylenedisulfotetramine solution, 100 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®