Merck
  • Direct aldol reactions catalyzed by intramolecularly folded prolinamide dendrons: dendrimer effects on stereoselectivity.
Direct aldol reactions catalyzed by intramolecularly folded prolinamide dendrons: dendrimer effects on stereoselectivity.
Chemical communications (Cambridge, England) (2009-07-10)
Kazuhiko Mitsui, Sarah A Hyatt, Daniel A Turner, Christopher M Hadad, Jon R Parquette
ABSTRACT
Dendritic effects on both the enantioselectivity and diastereoselectivity of the direct aldol reaction were observed for pyridine-2,6-dicarboxamide dendrons terminated with L-prolinamides.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
L-Prolinamide, 98%