Merck
  • Distinct extracytoplasmic siderophore binding proteins recognize ferrioxamines and ferricoelichelin in Streptomyces coelicolor A3(2).
Distinct extracytoplasmic siderophore binding proteins recognize ferrioxamines and ferricoelichelin in Streptomyces coelicolor A3(2).
Biochemistry (2010-08-14)
Prakash Patel, Lijiang Song, Gregory L Challis
ABSTRACT
Under iron limitation, the Gram-positive bacterium Streptomyces coelicolor A3(2) excretes three siderophores of the hydroxamate type: desferrioxamine B, desferrioxamine E, and coelichelin. These sequester iron from insoluble ferric hydroxides, and the resulting ferric complexes are believed to be transported into the cell via siderophore-binding proteins (SBPs) associated with ATP-binding cassette (ABC) transporters. Previous studies indicated that some of the genes in the desferrioxamine (des) and coelichelin (cch) biosynthetic clusters encode ABC transporter components required for efficient uptake of ferrioxamine E and ferricoelichelin, respectively, and a third ABC transporter gene cluster (cdt), not associated with siderophore biosynthesis genes, was implicated in the import of ferrioxamine B. In this study, the putative SBPs associated with these three gene clusters, DesE, CchF, and CdtB, were recombinantly overproduced in Escherichia coli and purified to homogeneity, and their binding affinity for cognate siderophores and noncognate siderophores was examined using fluorescence and circular dichroism spectroscopy. DesE was found to bind all of the ferric-tris-hydroxamates tested except ferricoelichelin, while CchF was found to bind only ferricoelichelin efficiently, providing further evidence that the cch cluster is a complete siderophore biosynthesis-export-uptake gene cluster. The picture was more complicated for CdtB, because it was found to be unstable in solution but was found to bind both ferrioxamine B and ferricoelichelin with high affinity. This was surprising because the cch cluster was previously reported to be necessary for efficient ferricoelichelin uptake. The remarkable specificity of the DesE and CchF proteins for different ferric-tris-hydroxamates raises intriguing questions about the molecular basis of their substrate specificity.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Millipore
Ferrioxamine E from Streptomyces antibioticus, suitable for microbiology, ≥95%