Merck
  • Effects of three types of alpha-1 adrenoceptor blocker on lower urinary tract symptoms and sexual function in males with benign prostatic hyperplasia.
Effects of three types of alpha-1 adrenoceptor blocker on lower urinary tract symptoms and sexual function in males with benign prostatic hyperplasia.
International journal of urology : official journal of the Japanese Urological Association (2011-01-29)
Teruhiko Yokoyama, Ryoei Hara, Kazuhiko Fukumoto, Tomohiro Fujii, Yoshimasa Jo, Yoshiyuki Miyaji, Atsushi Nagai, Atsushi Sone
ABSTRACT
The aim of the present study was to explore the effects of three different types of alpha-1 adrenoceptor blockers (α1-blocker) on lower urinary tract symptoms (LUTS), erectile dysfunction (ED) and ejaculatory dysfunction (EjD) in patients with benign prostatic hyperplasia. A total of 136 male LUTS patients aged 50-80 years with International Prostate Symptom Score (IPSS) ≥8 were enrolled. They were divided into three groups. Group S received silodosin at 4 mg twice a day; group T received tamsulosin at 0.2 mg once a day; and group N received naftopidil at 50 mg once a day. Assessment included IPSS, quality of life indexes (QOL), International Index of Erectile Function (IIEF-5), an ejaculation questionnaire, Qmax and post-void residual urine volume (PVR). These parameters were recorded at baseline, and at 1 and 3 months after treatment had ended. Mean IPSS and Qmax significantly improved after treatment in all groups without any significant difference among them. As for the IIEF-5 score, only group N significantly improved at 1 and 3 months. After treatment, 2.6 and 2.4% of patients complained of a de novo reduced volume of ejaculation in both groups T and N, respectively. Ten out of 41 patients (24.4%) complained of a total absence of antegrade ejaculation in group S after treatment. All three types of α1-blockers provided an objective and subjective improvement of LUTS in the present study population. However, erectile function only improved in patients treated with naftopidil and a higher rate of EjD was observed in those receiving silodosin. Because of their variable effects, we should consider the sexual dimension when prescribing α1-blockers for LUTS.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Naftopidil hydrochloride hydrate, solid