Merck
Transketolase in Trypanosoma brucei.
Molecular and biochemical parasitology (2011-05-17)
Sabine A Stoffel, Vincent P Alibu, Jane Hubert, Charles Ebikeme, Jean-Charles Portais, Frédéric Bringaud, M Ernst Schweingruber, Michael P Barrett
ABSTRACT
A single copy gene, encoding a protein highly similar to transketolase from other systems, was identified in the Trypanosoma brucei genome. The gene was expressed in E. coli and the purified protein demonstrated transketolase activity with K(m) values of 0.2mM and 0.8mM respectively for xylulose 5-phosphate and ribose 5-phosphate. A peroxisomal targeting signal (PTS-1) present at the C-terminus of the protein suggested a glycosomal localisation. However, subcellular localisation experiments revealed that while the protein was present in glycosomes it was found mainly within the cytosol and thus has a dual localisation. Transketolase activity was absent from the long slender bloodstream form of the parasite and the protein was not detectable in this life cycle stage, with the RNA present only at low abundance, indicating a strong differential regulation, being present predominantly in the procyclic form. The gene was knocked out from procyclic T. brucei and transketolase activity was lost but no growth phenotype was evident in the null mutants. Metabolite profiling to compare wild type and TKT null mutants revealed substantial increases in transketolase substrate metabolites coupled to loss of sedoheptulose 7-phosphate, a principal product of the transketolase reaction.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
D-Ribose 5-phosphate barium salt hexahydrate, ≥99.0% (TLC)