Merck
  • Cloning and functional characterization of a caffeic acid O-methyltransferase from Trigonella foenum-graecum L.
Cloning and functional characterization of a caffeic acid O-methyltransferase from Trigonella foenum-graecum L.
Molecular biology reports (2011-05-24)
Jian-Chun Qin, Ya-Mei Zhang, Chen-Yong Lang, Yan-Hua Yao, Hong-Yu Pan, Xiang Li
ABSTRACT
A cDNA encoding an O-methyltransferase (namely FGCOMT1) was identified from the medicinal plant Trigonella foenum-graecum L. The FGCOMT1 enzyme is a functional caffeic acid O-methyltransferase (COMT) and is localized in the cytosol. Kinetic analysis indicated that FGCOMT1 protein exhibited the highest catalyzing efficiency towards 5-hydroxy ferulic acid and caffeic acid as substrates, but did not possess the abilities to methylate either quercetin or tricetin in vitro. Furthermore, transformation of Arabidopsis loss-of-function Atomt1 mutant with a FGCOMT1 cDNA partially complements accumulation of sinapoyl derivatives but did not function to produce the major methylated flavonol isorhamnetin in seeds. The results from this study indicated that FGCOMT1 is a COMT with substrate preference to monomeric lignin precursors but is not involved in the flavonoid methylation in T. foenum-graecum L.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
3,4-Dihydroxy-5-methoxycinnamic acid, ≥95.0% (HPLC)