Merck
  • Two G-protein-coupled-receptor candidates, Cand2 and Cand7, are involved in Arabidopsis root growth mediated by the bacterial quorum-sensing signals N-acyl-homoserine lactones.
Two G-protein-coupled-receptor candidates, Cand2 and Cand7, are involved in Arabidopsis root growth mediated by the bacterial quorum-sensing signals N-acyl-homoserine lactones.
Biochemical and biophysical research communications (2011-12-31)
Guoping Jin, Fang Liu, Hong Ma, Shaoyan Hao, Qian Zhao, Zirui Bian, Zhenhua Jia, Shuishan Song
ABSTRACT
Many Gram-negative bacteria use N-acyl-homoserine lactones (AHLs) as quorum sensing (QS) signaling molecules to coordinate their group behavior. Recently, it was shown that plants can perceive and respond to these bacterial AHLs. However, little is known about the molecular mechanism underlying the response of plants to bacterial QS signals. In this study, we show that the promotion of root elongation in wild type Arabidopsis thaliana induced by the AHLs N-3-oxo-hexanoyl-homoserine lactone (3OC6-HSL) or N-3-oxo-octanoyl-homoserine lactone (3OC8-HSL) was completely abolished in plants with loss-of-function mutations in two candidate G-protein Coupled Receptors (GPCRs), Cand2 and Cand7. Furthermore, real-time PCR analysis revealed that the expression levels of Cand2 and Cand7 were elevated in plants treated with 3OC6-HSL or 3OC8-HSL. These results suggest that Cand2 and Cand7 are involved in the regulation of root growth by bacterial AHLs and that GPCRs play a role in mediating interactions between plants and microbes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
N-(β-Ketocaproyl)-DL-homoserine lactone, analytical standard