Merck
  • Enantio- and diastereocontrolled conversion of chiral epoxides to trans-cyclopropane carboxylates: application to the synthesis of cascarillic acid, grenadamide and L-(-)-CCG-II.
Enantio- and diastereocontrolled conversion of chiral epoxides to trans-cyclopropane carboxylates: application to the synthesis of cascarillic acid, grenadamide and L-(-)-CCG-II.
Organic & biomolecular chemistry (2012-07-27)
Pradeep Kumar, Abhishek Dubey, Anand Harbindu
ABSTRACT
An efficient high yielding improved method for the enantio- and diastereoselective cyclopropanation of chiral epoxides using triethylphosphonoacetate and base (Wadsworth-Emmons cyclopropanation) is reported. The utility of this protocol is illustrated by concise and practical synthesis of cascarillic acid, grenadamide and L-(-)-CCG-II, a cyclopropane containing natural products.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
(2S,3R,4S)-α-(Carboxycyclopropyl)glycine, solid