Merck
  • Blocking bimolecular activation pathways leads to different regioselectivity in metal-organic framework catalysis.
Blocking bimolecular activation pathways leads to different regioselectivity in metal-organic framework catalysis.
Chemical communications (Cambridge, England) (2012-07-28)
Teng Zhang, Feijie Song, Wenbin Lin
ABSTRACT
Chiral metal-organic frameworks built from a Co-salen derived dicarboxylate bridging ligand exhibited different regioselectivities from the corresponding homogeneous catalyst by shutting down bimolecular activation pathways.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N,N′-Bis(salicylidene)ethylenediamine, 98%