Merck
  • Naftopidil induces apoptosis in malignant mesothelioma cell lines independently of α1-adrenoceptor blocking.
Naftopidil induces apoptosis in malignant mesothelioma cell lines independently of α1-adrenoceptor blocking.
Anticancer research (2013-03-14)
Eriko Masachika, Takeshi Kanno, Takashi Nakano, Akinobu Gotoh, Tomoyuki Nishizaki
ABSTRACT
Naftopidil, an α1-adrenoceptor blocker, has been clinically used for the treatment of benign prostate hyperplasia and hypertension. Emerging evidence has shown that naftopidil exhibits an antitumor effect on a variety of cancer types including prostate cancer. The aim of the present study was to investigate naftopidil-induced apoptosis in human malignant mesothelioma cells and to shed light on the underlying mechanism. 3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide (MTT) assay, terminal deoxynucleotidyl transferase-mediated dUTP nick-end labeling (TUNEL) staining, western blotting, and enzymatic assay of caspase-3, -8, and -9 activities were carried out on human malignant mesothelioma cell lines NCI-H28, NCI-H2052, NCI-H2452, and MSTO-211H cells. To knock-down α1D-adrenoceptor, siRNA to silence human α1D-adrenoceptor-targeted gene was constructed and transfected into cells. Naftopidil induced apoptosis in all the investigated malignant mesothelioma cells, and a similar effect was obtained with prazosin, another α1-adrenoceptor blocker. α1-Adrenoceptor is linked to Gq/11 protein involving activation of protein kinase C (PKC). Naftopidil-induced reduction in cell viability was inhibited by GF109203X, while prazosin-induced in cell viability was less affected. Knocking-down α1D-adrenoceptor promoted malignant mesothelioma cell proliferation. Both naftopidil and prazosin activated caspase-3 and -8 in all the investigated malignant mesothelioma cells. Naftopidil, as well as prazosin, has the potential to induce apoptosis in malignant mesothelioma cells by activating caspase-8 and the effector caspase-3, regardless of α1-adrenoceptor blocking.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Naftopidil hydrochloride hydrate, solid