Merck
Permeability of the mitochondrial outer membrane to organic cations.
Biochimica et biophysica acta (1989-06-06)
C A Mannella, Q Wang
ABSTRACT
Inhibition of mitochondrial respiration by hydrophobic fluorescent dyes (Rhodamine 6G, Safranine O, Pyronine B) is much less potentiated by digitonin-lysis of the outer membrane than that by polyamines or adriamycin. This situation may be explained by impermeability of the anion-selective channels in the outer mitochondrial membrane to large cations and by the ability of hydrophobic (but not polar or amphipathic) ions to directly permeate lipid bilayers.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Pyronin B, Dye content ≥30 %