- Eimeria stiedai in rabbits: the demonstration of responses to chemotherapy.
Eimeria stiedai in rabbits: the demonstration of responses to chemotherapy.
Research in veterinary science (1983-01-01)
L P Joyner, J Catchpole, S Berrett
PMID6132429
ABSTRACT
The activity of sulphaquinoxaline, robenidine, methyl benzoquate, clopidol and a mixture of methyl benzoquate and clopidol (Lerbek; Dow), was studied in rabbits infected with hepatic coccidiosis due to Eimeria stiedai. Growth inhibition, oocyst production and the activity in the serum of glutamate dehydrogenase and gamma glutamyltransferase were studied as indicators of parasite development. Only sulphaquinoxaline and Lerbek gave satisfactory control of this parasite. The latter formulation was more effective than either of its constituents used alone.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto